Anti-ZIC4 Mouse Monoclonal Antibody [clone: 1A9]
USBI135571
- Antibody type:Primary
- Antigen name:Zic family member 4
- Antigen symbol:ZIC4
- Clonality:Monoclonal
- Clone:1A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:249 to 320
- Storage buffer:PBS, pH 7,2.
- Sequence:SDRKKHSHVHTSDKPYTCKVRGCDKCYTHPSSLRKHMKVHGRSPPPSSGYDSATPSALVSPSSDCGHKSQV
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml
Type: Primary
Antigen: ZIC4 (Zic family member 4)
Clonality: monoclonal
Clone: 1A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human