Anti-VAMP1 Mouse Monoclonal Antibody [clone: 5A4]
USBI135197
- Antibody type:Primary
- Antigen name:Vesicle-Associated Membrane Protein 1
- Antigen symbol:VAMP1
- Clonality:Monoclonal
- Clone:5A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a lambda
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:28 to 97
- Storage buffer:PBS, pH 7,2.
- Sequence:MTSNRRLQQTQAQVEEVVDIIRVNVDKVLERDQKLSELDDRADALQAGASQFESSAAKLKRKYWWKNCK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1. 5 ?g/ml
Type: Primary
Antigen: VAMP1 (vesicle-associated membrane protein 1 (synaptobrevin 1))
Clonality: monoclonal
Clone: 5A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Lambda
Reactivity: Human