Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Tetratricopeptide Repeat Protein 18 Mouse Polyclonal Antibody
Catalog # USBI134851
Supplier:  US Biological
Anti-Tetratricopeptide Repeat Protein 18 Mouse Polyclonal Antibody
Catalog # USBI134851
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tetratricopeptide Repeat Protein 18
  • Antigen symbol:
    TTC18
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    1 to 1121
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Tetratricopeptide Repeat Protein 18
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human