Anti-TCF19 Mouse Monoclonal Antibody [clone: 6D8]
Catalog # USBI134289
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor 19
- Antigen symbol:TCF19
- Clonality:Monoclonal
- Clone:6D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:17 to 103
- Storage buffer:PBS, pH 7,2.
- Sequence:DLYTFHPPAGVGCTYRLGHRADLCDVALRPQQEPGLISGIHAELHAEPRGDDWRVSLEDHSSQGTLVNNVRLPRGHRLELSDGDLL
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml
Type: Primary
Antigen: TCF19 (transcription factor 19)
Clonality: monoclonal
Clone: 6D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human