Anti-SLC7A7 Mouse Monoclonal Antibody [clone: 3B10]
USBI133516
- Antibody type:Primary
- Antigen name:solute carrier family 7 (amino acid transporter light chain, y+L system), member 7
- Antigen symbol:Solute carrier family 7, member 7
- Clonality:Monoclonal
- Clone:3B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:462 to 512
- Storage buffer:PBS, pH 7,2.
- Sequence:RVPEHKRPLYLRRIVGSATRYLQVLCMSVAAEMDLEDGGEMPKQRDPKSN
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SLC7A7 (solute carrier family 7 (amino acid transporter light chain, y+L system), member 7)
Clonality: monoclonal
Clone: 3B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human