Anti-SLC6A19 Mouse Monoclonal Antibody [clone: 4F11]
Catalog # USBI133508
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 6 (neutral amino acid transporter), member 19
- Antigen symbol:Solute carrier family 6, member 19
- Clonality:Monoclonal
- Clone:4F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:326 to 415
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC6A19 (solute carrier family 6 (neutral amino acid transporter), member 19)
Clonality: monoclonal
Clone: 4F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human