Anti-RING Finger Protein 20 Rabbit Polyclonal Antibody
USBI132684
- Antibody type:Primary
- Antigen name:Ring Finger Protein 20
- Antigen symbol:RNF20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Format:Serum
- Form:liquid
- Amino acid number:1 to 975
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: RING Finger Protein 20
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human