Anti-PSMD2 Mouse Monoclonal Antibody [clone: 1B4]
USBI131975
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) 26S subunit, non-ATPase 2
- Antigen symbol:PSMD2
- Clonality:Monoclonal
- Clone:1B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: PSMD2 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 2)
Clonality: monoclonal
Clone: 1B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human