Anti-POLR2H Mouse Monoclonal Antibody [clone: 3G6-1A4]
Catalog # USBI131589
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (RNA) II (DNA directed) polypeptide H
- Antigen symbol:POLR2H
- Clonality:Monoclonal
- Clone:3G6-1A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MAGILFEDIFDVKDIDPEGKKFDRVSRLHCESESFKMDLILDVNIQIYPVDLGDKFRLVIASTLYEDGTLDDGEYNPTDDRPSRADQFEYVMYGKVYRIEGDETSTEAATRLSAYVSYGGLLMRLQGDANNLHGFEVDSRVYLLMKKLAF
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: POLR2H (polymerase (RNA) II (DNA directed) polypeptide H)
Clonality: monoclonal
Clone: 3G6-1A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human