Anti-PEX11A Mouse Monoclonal Antibody [clone: 3B1]
Catalog # USBI131165
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:peroxisomal biogenesis factor 11 alpha
- Antigen symbol:PEX11A
- Clonality:Monoclonal
- Clone:3B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:1 to 94
- Storage buffer:PBS, pH 7,2.
- Sequence:MDAFTRFTNQTQGRDRLFRATQYTCMLLRYLLEPKAGKEKVVMKLKKLESSVSTGRKWFRLGNVVHAIQATEQSIHATDLVPRLCLTLANLNR
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PEX11A (peroxisomal biogenesis factor 11 alpha)
Clonality: monoclonal
Clone: 3B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human