Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PDIA5 Mouse Monoclonal Antibody [clone: 3A3]
Anti-PDIA5 Mouse Monoclonal Antibody [clone: 3A3]
  USBI131085
Anti-PDIA5 Mouse Monoclonal Antibody [clone: 3A3]
  USBI131085

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein Disulfide Isomerase Family A, Member 5
  • Antigen symbol:
    PDIA5
  • Clonality:
    Monoclonal
  • Clone:
    3A3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    31 to 131
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    ERISDPKDLKKLLRTRNNVLVLYSKSEVAAENHLRLLSTVAQAVKGQGTICWVDCGDAESRKLCKKMKVDLSPKDKKVELFHYQDGAFHTEYNRAVTFKS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: PDIA5 (protein disulfide isomerase family A, member 5)
Clonality: monoclonal
Clone: 3A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human