Anti-NETO1 Mouse Polyclonal Antibody
USBI130293
- Antibody type:Primary
- Antigen name:Neuropilin (NRP) and tolloid (TLL)-like 1
- Antigen symbol:NETO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: NETO1 (neuropilin (NRP) and tolloid (TLL)-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human