Anti-NDUFV1 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # USBI130247
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa
- Antigen symbol:NDUFV1
- Clonality:Monoclonal
- Clone:4A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:365 to 464
- Storage buffer:PBS, pH 7,2.
- Sequence:KAIARLIEFYKHESCGQCTPCREGVDWMNKVMARFVRGDARPAEIDSLWEISKQIEGHTICALGDGAAWPVQGLIRHFRPELEERMQRFAQQHQARQAAS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NDUFV1 (NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa)
Clonality: monoclonal
Clone: 4A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human