Anti-MFN2 Mouse Monoclonal Antibody [clone: 4H8]
Catalog # USBI129610
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:mitofusin 2
- Antigen symbol:MFN2
- Clonality:Monoclonal
- Clone:4H8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Amino acid number:661 to 757
- Storage buffer:PBS, pH 7,2.
- Sequence:FKRQFVEHASEKLQLVISYTGSNCSHQVQQELSGTFAHLCQQVDVTRENLEQEIAAMNKKIEVLDSLQSKAKLLRNKAGWLDSELNMFTHQYLQPSR
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: MFN2 (mitofusin 2)
Clonality: monoclonal
Clone: 4H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat