Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LDHB Mouse Monoclonal Antibody [clone: 2H6]
Anti-LDHB Mouse Monoclonal Antibody [clone: 2H6]
Catalog # USBI129039
Supplier:  US Biological
Anti-LDHB Mouse Monoclonal Antibody [clone: 2H6]
Catalog # USBI129039
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lactate dehydrogenase B
  • Antigen symbol:
    LDHB
  • Clonality:
    Monoclonal
  • Clone:
    2H6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MATLKEKLIAPVAEEEATVPNNKITVVGVGQVGMACAISILGKSLADELALVDVLEDKLKGEMMDLQHGSLFLQTPKIVADKDYSVTANSKIVVVTAGVRQQEGESRLNLVQRNVNVFKFIIPQIVKYSPDCIIIVVSNPVDILTYVTWKLSGLPKHRVIGSGCNLDSARFRYLMAEKLGIHPSSCHGWILGEHGDSSVAVWSGVNVAGVSLQELNPEMGTDNDSENWKEVHKMVVESAYEVIKLKGYTNWAIGLSVADLIESMLKNLSRIHPVSTMVKGMYGIENEVFLSLPCILNARGLTSDINQKLKDDEVAQLKKSADTLWDIQKDLKDL
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: LDHB (lactate dehydrogenase B)
Clonality: monoclonal
Clone: 2H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human