Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IL27 Mouse Monoclonal Antibody [clone: 3F12]
  USBI128456
Anti-IL27 Mouse Monoclonal Antibody [clone: 3F12]
  USBI128456

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 27
  • Antigen symbol:
    IL27
  • Clonality:
    Monoclonal
  • Clone:
    3F12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    177 to 244
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: IL27 (interleukin 27)
Clonality: monoclonal
Clone: 3F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human