Anti-EDIL3 Mouse Monoclonal Antibody [clone: 4C9]
Catalog # USBI126160
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:EGF-like repeats and discoidin I-like domains 3
- Antigen symbol:EDIL3
- Clonality:Monoclonal
- Clone:4C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:101 to 199
- Storage buffer:PBS, pH 7,2.
- Sequence:QHNINECEVEPCKNGGICTDLVANYSCECPGEFMGRNCQYKCSGPLGIEGGIISNQQITASSTHRALFGLQKWYPYYARLNKK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: EDIL3 (EGF-like repeats and discoidin I-like domains 3)
Clonality: monoclonal
Clone: 4C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse