Anti-PPIE Mouse Monoclonal Antibody [clone: 2F5]
Catalog # USBI125546
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:peptidylprolyl isomerase E (cyclophilin E)
- Antigen symbol:PPIE
- Clonality:Monoclonal
- Clone:2F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:2 to 100
- Storage buffer:PBS, pH 7,2.
- Sequence:ATTKRVLYVGGLAEEVDDKVLHAAFIPFGDITDIQIPLDYETEKHRGFAFVEFELAEDAAAAIDNMNESELFGRTIRVNLAKPMRIKEGSSRPVWSDDD
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: PPIE (peptidylprolyl isomerase E (cyclophilin E))
Clonality: monoclonal
Clone: 2F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat