Anti-CDSN Mouse Monoclonal Antibody [clone: 6F11]
USBI125236
- Antibody type:Primary
- Antigen name:Corneodesmosin
- Antigen symbol:CDSN
- Clonality:Monoclonal
- Clone:6F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:306 to 355
- Storage buffer:PBS, pH 7,2.
- Sequence:YLVPGMTYSKGKIYPVGYFTKENPVKGSPGVPSFAAGPPISEGKYFSSNP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CDSN (corneodesmosin)
Clonality: monoclonal
Clone: 6F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human