Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CES3 Mouse Polyclonal Antibody
  USBI124882
Anti-CES3 Mouse Polyclonal Antibody
  USBI124882

 

  • Antibody type:
    Primary
  • Antigen name:
    Carboxylesterase 3
  • Antigen symbol:
    CES3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: CES3 (carboxylesterase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human