Anti-ANKRD17 Mouse Monoclonal Antibody [clone: 2B8]
Catalog # USBI123339
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Ankyrin repeat domain 17
- Antigen symbol:ANKRD17
- Clonality:Monoclonal
- Clone:2B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:2501 to 2604
- Storage buffer:PBS, pH 7,2.
- Sequence:ERDSTGIVTPSGTFHQHVPAGYMDFPKVGGMPFSVYGNAMIPPVAPIPDGAGGPIFNGPHAADPSWNSLIKMVSSSTENNGPQTVWTGPWAPHMNSVHMNQLG*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: ANKRD17 (ankyrin repeat domain 17)
Clonality: monoclonal
Clone: 2B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human