Anti-SLC7A6 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBI043960-PE
- Antibody type:Primary
- Antigen name:solute carrier family 7 (amino acid transporter light chain, y+L system), member 6
- Antigen symbol:Solute carrier family 7, member 6
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SLC7A6 (solute carrier family 7 (amino acid transporter light chain, y+L system), member 6)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human