Anti-SLC16A10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI041801-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 16 (aromatic amino acid transporter), member 10
- Antigen symbol:Solute carrier family 16, member 10
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:301 to 329
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SLC16A10 (solute carrier family 16 (aromatic amino acid transporter), member 10)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human