Anti-RGS9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI041018-FITC
Supplier: US Biological
Specifications
- Druh protilátky:Primární
- Název antigenu:Regulator of G-protein signaling 9
- Symbol antigenu:RGS9
- Klonovalita:Polyclonal
- Spojení:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human
- Western blot:Yes
- Epitop:N-terminal
- Formulář:liquid
- Počet aminokyselin:148 to 178
- Skladovací pufr:PBS, pH 7,2, contains ~0,09% sodium azide
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Testované aplikace:Fluoresence ELISA
- Čištění:purified by Protein A chromatography
- Rozměry protilátky:200 µl
- Bal.:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: RGS9 (regulator of G-protein signaling 9)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human