Anti-PI3KC3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBI039979-PE
- Antibody type:Primary
- Antigen symbol:PI3KC3
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:19 to 49
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot1:50-100; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: PI3KC3
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human