Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PI3KC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-PI3KC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI039979-FITC
Supplier:  US Biological
undefined
Anti-PI3KC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI039979-FITC
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PI3KC3
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    19 to 49
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot1:50-100; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PI3KC3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human