Anti-PARP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI039671-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Poly (ADP-ribose) polymerase 1
- Antigen symbol:PARP1
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Modification:phospho-Ser373
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Dot Blot, Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Dot blot 1:500
Do not freeze!
Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: