Order Entry
Czech Republic
ContactUsLinkComponent
Anti-NDUAB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-NDUAB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalogové číslo USBI038853-FITC
Dodavatel:  US Biological
undefined
Anti-NDUAB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalogové číslo USBI038853-FITC
Dodavatel:  US Biological

Specifikace

  • Antibody type:
    Primary
  • Antigen symbol:
    NDUAB
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Form:
    liquid
  • Amino acid number:
    71 to 100
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifikace

O tomto produktu

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NDUAB
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human