Anti-JUN Rabbit Polyclonal Antibody
Catalog # USBI037228
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:jun proto-oncogene
- Antigen symbol:JUN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Modification:phospho-Thr289
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Dot blot 1:500
Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse