Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ERBB2 Rabbit Polyclonal Antibody
Anti-ERBB2 Rabbit Polyclonal Antibody
Catalog # USBI035181
Supplier:  US Biological
undefined
Anti-ERBB2 Rabbit Polyclonal Antibody
Catalog # USBI035181
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
  • Antigen symbol:
    ERBB2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    liquid
  • Modification:
    phospho-Tyr1127
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Dot blot 1:500

Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human