Anti-AES Rabbit Polyclonal Antibody
Catalog # USBI031670
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:amino-terminal enhancer of split
- Antigen symbol:AES
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:155 to 184
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: AES (amino-terminal enhancer of split)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human