Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Tar DNA Binding Protein 43 Mouse Monoclonal Antibody [clone: 13B862]
  USBI031432
Anti-Tar DNA Binding Protein 43 Mouse Monoclonal Antibody [clone: 13B862]
  USBI031432

 

  • Antibody type:
    Primary
  • Antigen name:
    TAR DNA-binding protein 43
  • Antigen symbol:
    TDP43
  • Clonality:
    Monoclonal
  • Clone:
    13B862
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:5000

Type: Primary
Antigen: Tar DNA Binding Protein 43
Clonality: Monoclonal
Clone: 13B862
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Pig, Rat