Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PRKAA2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 206CT5.4.2]
Anti-PRKAA2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 206CT5.4.2]
Catalog # USBI031207-PE
Supplier:  US Biological
undefined
Anti-PRKAA2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 206CT5.4.2]
Catalog # USBI031207-PE
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein kinase, AMP-activated, alpha 2 catalytic subunit
  • Antigen symbol:
    PRKAA2
  • Clonality:
    Monoclonal
  • Clone:
    206CT5.4.2
  • Conjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze! Light sensitive

Type: Primary
Antigen: PRKAA2 (protein kinase, AMP-activated, alpha 2 catalytic subunit)
Clonality: Monoclonal
Clone: 206CT5.4.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human