Anti-PRKAA2 Mouse Monoclonal Antibody [clone: 206CT5.4.2]
Catalog # USBI031206
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 2 catalytic subunit
- Antigen symbol:PRKAA2
- Clonality:Monoclonal
- Clone:206CT5.4.2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-2000
Type: Primary
Antigen: PRKAA2 (protein kinase, AMP-activated, alpha 2 catalytic subunit)
Clonality: Monoclonal
Clone: 206CT5.4.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse