Anti-MAP Kinase 15 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 168CT10.6.6]
USBI031100-FITC
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 15
- Antigen symbol:MAPK15
- Clonality:Monoclonal
- Clone:168CT10.6.6
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Type: Primary
Antigen: MAP Kinase 15
Clonality: Monoclonal
Clone: 168CT10.6.6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human