Anti-CLIC5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 16CT34.4.8]
USBI030877-FITC
- Antibody type:Primary
- Antigen name:chloride intracellular channel 5
- Antigen symbol:CLIC5
- Clonality:Monoclonal
- Clone:16CT34.4.8
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:50-100
Type: Primary
Antigen: CLIC5 (chloride intracellular channel 5)
Clonality: Monoclonal
Clone: 16CT34.4.8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse