Anti-PMAIP1 Rabbit Polyclonal Antibody
Catalog # USBI029562
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:phorbol-12-myristate-13-acetate-induced Protein 1
- Antigen symbol:PMAIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 1:500
Type: Primary
Antigen: PMAIP1 (phorbol-12-myristate-13-acetate-induced protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human