Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Interleukin-10 Rabbit Polyclonal Antibody
  USBI029517
Anti-Interleukin-10 Rabbit Polyclonal Antibody
  USBI029517

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin-10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 0. 01 ?g/ml

Type: Primary
Antigen: Interleukin-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse