Anti-FGF21 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI029150
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast growth factor 21
- Antigen symbol:FGF21
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Supplied as a liquid in PBS, pH 7.4, 0.03% Proclin 300, 50% glycerol. Labeled with Fluorescein isothiocyanate (FITC).
- Antigen synonyms:FGF-21|UNQ3115/PRO10196
- Storage buffer:0,01M PBS, PH 7.4, 0,03% Procl300, 50% glycerol.
- Storage temperature:–20 °C
- Shipping temperature:20
- Immunogen:Recombinant protein corresponding to human Fibroblast growth factor 21.
- Purification:Purified by caprylic acid ammonium sulfate precipitation.
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FGF21 (fibroblast growth factor 21)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human