Anti-SELL Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MEL-14]
ROCK200-508-N87
- Antibody type:Primary
- Antigen name:Selectin L
- Antigen symbol:SELL
- Clonality:Monoclonal
- Clone:MEL-14
- Conjugation:RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:20343
- Antigen synonyms:Lyam-1|Lnhr|Lyam1|Ly-m22|Ly-22|LECAM-1|CD62L|AI528707|L-selectin
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,2 mg/ml Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD62L Antibody (Monoclonal) was produced by repeated immunizations with C3H/eb mouse B lymphoma 38C-13.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:200 µg
- Pk:200 µG
Anti-CD62L is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: MEL-14
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse