Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SELL Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: DREG-56]
Anti-SELL Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: DREG-56]
Catalog # ROCK200-308-N72
undefined
Anti-SELL Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: DREG-56]
Catalog # ROCK200-308-N72

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Selectin L
  • Antigen symbol:
    SELL
  • Clonality:
    Monoclonal
  • Clone:
    DREG-56
  • Conjugation:
    RPE (R-Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    6402
  • Antigen synonyms:
    LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,2% BSA (w/v)
  • Storage temperature:
    Cool
  • Concentration:
    Sufficient to run approximately 100 tests
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-CD62L Antibody (Monoclonal) was produced by repeated immunizations with CD62L antigen.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

Anti-CD62L is useful for Flow Cytometry and Western Blot. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions:
Do not freeze! Light sensitive!

Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: DREG-56
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human