Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG (H+L) Donkey Polyclonal Antibody (Cy5.5®)
Anti-IgG (H+L) Donkey Polyclonal Antibody (Cy5.5®)
Catalog # ROCK609-713-123
Anti-IgG (H+L) Donkey Polyclonal Antibody (Cy5.5®)
Catalog # ROCK609-713-123

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Donkey
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2", Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Human IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Human