Anti-CDKN2A (cyclin-dependent Kinase Inhibitor 2A) Rabbit Polyclonal Antibody
Catalog # ROCK100-401-170
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 2A
- Antigen symbol:CDKN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:1029
- Antigen synonyms:CDKN2|MLM|INK4|MTS-1|P16|P16INK4|P19|P16-INK4A|P16INK4A|INK4A|P19ARF|CDK4I|CMM2|P14ARF|P14|ARF|MTS1|TP16
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CDKN2A (cyclin-dependent Kinase Inhibitor 2A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human