Anti-Insulin Growth Factor-1 Receptor (IGF-1R) Mouse Monoclonal Antibody
Catalog # QEDB35102
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Insulin Growth Factor-1 Receptor
- Antigen symbol:IGF-1R
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Insulin Growth Factor-1 Receptor (IGF-1R)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human