Anti-BCAS1 / NABC1 Rabbit Polyclonal Antibody
Catalog # QEDB34017
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen symbol:BCAS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:NABC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: BCAS1 / NABC1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human