Anti-Fibroblast Growth Factor Receptor 1 alpha Mouse monoclonal antibody
Catalog # QEDB30105
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast growth factor receptor 1A
- Antigen symbol:FGFR1A
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Type: Primary
Antigen: Fibroblast Growth Factor Receptor 1 alpha
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human