Anti-FBXO6 Rabbit Polyclonal Antibody
Katalogové číslo QEDB28119
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:F-box protein 6
- Antigen symbol:FBXO6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FBXO6 (F-box protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat