Order Entry
Czech Republic
ContactUsLinkComponent
Anti-STRADB Rabbit Polyclonal Antibody
Anti-STRADB Rabbit Polyclonal Antibody
Catalog # QEDB28107
Supplier:  QED Bioscience
Anti-STRADB Rabbit Polyclonal Antibody
Catalog # QEDB28107
Supplier:  QED Bioscience

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    STE20-related kinase adaptor beta
  • Antigen symbol:
    STRADB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: STRADB (STE20-related kinase adaptor beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse