Anti-SLC2A10 Rabbit Polyclonal Antibody
Catalog # QEDB28103
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 2 (facilitated glucose transporter), member 10
- Antigen symbol:SLC2A10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC2A10 (solute carrier family 2 (facilitated glucose transporter), member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human