Anti-Staphylococcus aureus Mouse Monoclonal Antibody
Catalog # QEDB15702
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen symbol:Staphylococcus aureus
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Staphylococcus aureus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: